α,α-Difluoroanisole 98%

Supplier: Apollo Scientific
Warning

Synonyms: (Difluoromethoxy)benzene, alpha,alpha-Difluoroanisole

PC4117-100G
APOSPC4117-100GEA 112 EUR
APOSPC4117-100G
α,α-Difluoroanisole 98%
α,α-Difluoroanisole
Formula: C₇H₆F₂O
MW: 144,1200 g/mol
Boiling Pt: 98…100 °C
Density: 1,2 g/cm³ (25 °C)
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00068184
CAS Number: 458-92-4
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR