3-Fluoro-4-pyridineboronic acid pinacol ester 95%

Supplier: Apollo Scientific

PC412004-10G PC412004-25G PC412004-5G
APOSPC412004-10GEA 125 EUR
APOSPC412004-10G APOSPC412004-25G APOSPC412004-5G
3-Fluoro-4-pyridineboronic acid pinacol ester 95%
3-Fluoro-4-pyridineboronic acid pinacol ester
Formula: C₁₁H₁₅BFNO₂
MW: 223,05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06798251
CAS Number: 458532-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR