4-Fluoro-3-phenoxybenzaldehyde 98%
Supplier: Apollo Scientific
|
Warning
|
PC4157-250G
PC4157-50G
PC4157-5G
APOSPC4157-250GEA
464
EUR
APOSPC4157-250G
APOSPC4157-50G
APOSPC4157-5G
4-Fluoro-3-phenoxybenzaldehyde 98%
4-Fluoro-3-phenoxybenzaldehyde
Formula:
C₁₃H₉FO₂ Boiling Pt: 310 °C Density: 1,209 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD01318148 CAS Number: 68359-57-9 EINECS: 269-856-2 UN: 3082 ADR: 9+EHS,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...