4-Fluorophenyl isocyanate 98%

Supplier: Apollo Scientific
Danger

Synonyms: 4-Fluorophenylisocyanate, 1-Fluoro-4-isocyanatobenzene

PC4166-100G PC4166-25G PC4166-5G
APOSPC4166-100GEA 127 EUR
APOSPC4166-100G APOSPC4166-25G APOSPC4166-5G
4-Fluorophenyl isocyanate 98%
4-Fluorophenyl isocyanate

  • May form small amount of sediment.

Formula: C₇H₄FNO
Boiling Pt: 55 °C/8mmHg
Density: 1,206 g/cm³ (25 °C)
Flash Pt: 52 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002023
CAS Number: 1195-45-5
EINECS: 214-799-0
UN: 3080
ADR: 6.1+3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR