1-(4-Fluorophenyl)pyrrole 97%

Supplier: Apollo Scientific

Synonyms: N-(4-Fluorophenyl)pyrrole, 1-(4-Fluorophenyl)-1H-pyrrole

PC4174M-1G PC4174M-5G
APOSPC4174M-1GEA 94.5 EUR
APOSPC4174M-1G APOSPC4174M-5G
1-(4-Fluorophenyl)pyrrole 97%
1-(4-Fluorophenyl)pyrrole
Formula: C₁₀H₈FN
MW: 161,18 g/mol
Melting Pt: 52 °C
Storage Temperature: Ambient
MDL Number: MFCD00047066
CAS Number: 81329-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR