4-(2-Fluorophenyl)-3-thiosemicarbazide 97%

Supplier: Apollo Scientific
Danger

Synonyms: N-(2-Fluorophenyl)hydrazinecarbothioamide

PC4175J-1G PC4175J-2.5G PC4175J-250MG
APOSPC4175J-1GEA 16.1 EUR
APOSPC4175J-1G APOSPC4175J-2.5G APOSPC4175J-250MG
4-(2-Fluorophenyl)-3-thiosemicarbazide 97%
4-(2-Fluorophenyl)-3-thiosemicarbazide
Formula: C₇H₈FN₃S
MW: 185,22 g/mol
Melting Pt: 154…157 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00041269
CAS Number: 38985-72-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR