4-Hydroxy-2-(trifluoromethyl)pyrimidine 97%

Supplier: Apollo Scientific
Warning

Synonyms: 2-(Trifluoromethyl)pyrimidin-4-ol

PC4183-250MG PC4183-5G PC4183-25G PC4183-100G
APOSPC4183-250MGEA 0 EUR
APOSPC4183-250MG APOSPC4183-5G APOSPC4183-25G APOSPC4183-100G
4-Hydroxy-2-(trifluoromethyl)pyrimidine 97%
4-Hydroxy-2-(trifluoromethyl)pyrimidine
Formula: C₅H₃F₃N₂O
MW: 164,09 g/mol
Melting Pt: 168…171 °C
Storage Temperature: Ambient
MDL Number: MFCD00151833
CAS Number: 1546-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR