(+/-)-2-(Trifluoromethyl)piperazine 95%
Supplier: Apollo Scientific
Synonyms:
2-(Trifluoromethyl)piperazine
PC421203-100MG
PC421203-250MG
APOSPC421203-100MGEA
547
EUR
APOSPC421203-100MG
APOSPC421203-250MG
(+/-)-2-(Trifluoromethyl)piperazine 95%
(+/-)-2-(Trifluoromethyl)piperazine
Formula:
C₅H₉F₃N₂ MW: 154,13 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07373382 CAS Number: 131922-05-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...