2-Chloro-4-fluorobenzotrifluoride 98%
Supplier: Apollo Scientific
|
Warning
|
PC4268-5G
PC4268-1G
PC4268-25G
APOSPC4268-5GEA
24.8
EUR
APOSPC4268-5G
APOSPC4268-1G
APOSPC4268-25G
2-Chloro-4-fluorobenzotrifluoride 98%
2-Chloro-4-fluorobenzotrifluoride
Formula:
C₇H₃ClF₄ MW: 198,5453 g/mol Boiling Pt: 102…105 °C Melting Pt: –42 °C Density: 2,9 g/cm³ (25 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD03412197 CAS Number: 94444-58-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...