3-Bromo-4-fluorobenzoic acid 98%

Supplier: Apollo Scientific
Warning

PC4279-25G PC4279-5G
APOSPC4279-25GEA 58.2 EUR
APOSPC4279-25G APOSPC4279-5G
3-Bromo-4-fluorobenzoic acid 98%
3-Bromo-4-fluorobenzoic acid
Formula: C₇H₄BrFO₂
MW: 219,01 g/mol
Melting Pt: 138…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00042463
CAS Number: 1007-16-5
EINECS: 213-751-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR