2-(Difluoromethoxy)benzyl alcohol 98%
Supplier: Apollo Scientific
|
Warning
|
PC4303-5G
PC4303-25G
PC4303-100G
APOSPC4303-5GEA
37.7
EUR
APOSPC4303-5G
APOSPC4303-25G
APOSPC4303-100G
2-(Difluoromethoxy)benzyl alcohol 98%
2-(Difluoromethoxy)benzyl alcohol
Formula:
C₈H₈F₂O₂ MW: 174.15 g/mol Melting Pt: 42 °C Storage Temperature: Ambient |
MDL Number:
MFCD00236226 CAS Number: 72768-94-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...