3-Chloro-4-fluorobenzoyl chloride 95%
Supplier: Apollo Scientific
|
Danger
|
PC4308-25G
APOSPC4308-25GEA
95.7
EUR
APOSPC4308-25G
3-Chloro-4-fluorobenzoyl chloride 95%
3-Chloro-4-fluorobenzoyl chloride
Formula:
C₇H₃Cl₂FO Boiling Pt: 222 °C Density: 1,46 g/cm³ (25 °C) Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00236215 CAS Number: 65055-17-6 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...