2-Fluorotoluene 99%

Supplier: Apollo Scientific
Danger

Synonyms: 1-Fluoro-2-methylbenzene, o-Fluorotoluene

PC4310-1KG PC4310-250G PC4310-5KG
APOSPC4310-1KGEA 225 EUR
APOSPC4310-1KG APOSPC4310-250G APOSPC4310-5KG
2-Fluorotoluene 99%
2-Fluorotoluene
Formula: C₇H₇F
MW: 110.13 g/mol
Boiling Pt: 113…114 °C
Melting Pt: –62 °C
Density: 1.001 g/cm³ (25 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00000322
CAS Number: 95-52-3
EINECS: 202-428-5
UN: 2388
ADR: 3 II
Merck Index: 13,04207

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR