2-Fluoro-3-(trifluoromethyl)benzamide 98%

Supplier: Apollo Scientific

PC4371R-25G PC4371R-5G PC4371R-1G
APOSPC4371R-25GEA 139 EUR
APOSPC4371R-25G APOSPC4371R-5G APOSPC4371R-1G
2-Fluoro-3-(trifluoromethyl)benzamide 98%
2-Fluoro-3-(trifluoromethyl)benzamide
Formula: C₈H₅F₄NO
Melting Pt: 93…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00061146
CAS Number: 207853-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR