2-Fluoro-6-(trifluoromethyl)benzoyl chloride 98%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
2-Fluoro-6-(trifluoromethyl)benzene-1-carbonyl chloride, 2-Fluoro-6-(trifluoromethyl)benzoylchloride
PC4374P-25G
PC4374P-5G
PC4374P-1G
APOSPC4374P-25GEA
386
EUR
APOSPC4374P-25G
APOSPC4374P-5G
APOSPC4374P-1G
2-Fluoro-6-(trifluoromethyl)benzoyl chloride 98%
2-Fluoro-6-(trifluoromethyl)benzoyl chloride
Formula:
C₈H₃ClF₄O Boiling Pt: 194 °C Density: 1,465 g/cm³ (25 °C) Flash Pt: 82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00061160 CAS Number: 109227-12-5 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...