2-Fluoro-3-(trifluoromethyl)benzyl bromide 98%

Supplier: Apollo Scientific
Danger

PC4376E-5G PC4376E-1G PC4376E-25G
APOSPC4376E-5GEA 94.4 EUR
APOSPC4376E-5G APOSPC4376E-1G APOSPC4376E-25G
2-Fluoro-3-(trifluoromethyl)benzyl bromide 98%
2-Fluoro-3-(trifluoromethyl)benzyl bromide
Formula: C₈H₅BrF₄
MW: 257,03 g/mol
Melting Pt: 58…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00061172
CAS Number: 184970-25-0
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR