2-Fluoro-5-(trifluoromethyl)benzyl bromide 97%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
1-(Bromomethyl)-2-fluoro-5-(trifluoromethyl)benzene, 2-Fluoro-5-(trifluoromethyl)benzyl Bromide
PC4376G-1G
PC4376G-5G
PC4376G-25G
APOSPC4376G-1GEA
21.9
EUR
APOSPC4376G-1G
APOSPC4376G-5G
APOSPC4376G-25G
2-Fluoro-5-(trifluoromethyl)benzyl bromide 97%
2-Fluoro-5-(trifluoromethyl)benzyl bromide
Formula:
C₈H₅BrF₄ MW: 257,03 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00061174 CAS Number: 220239-69-0 UN: 1760 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...