3-Fluoro-DL-tyrosine 97%

Supplier: Apollo Scientific
Warning

Synonyms: H-DL-Tyr(3-F)-OH, 2-Amino-3-(3-fluoro-4-hydroxyphenyl)propanoic acid

PC4379-1G PC4379-250MG
APOSPC4379-1GEA 259 EUR
APOSPC4379-1G APOSPC4379-250MG
3-Fluoro-DL-tyrosine 97%
3-Fluoro-DL-tyrosine
Formula: C₉H₁₀FNO₃
MW: 199,18 g/mol
Melting Pt: 280 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00063075
CAS Number: 403-90-7
EINECS: 206-964-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR