5-Fluorouracil 99%

Supplier: Apollo Scientific
Danger

Synonyms: Fluorouracil, 5-Fluoropyrimidine-2,4(1H,3H)-dione, 5-FU

PC4380-100G PC4380-25G
APOSPC4380-100GEA 48.1 EUR
APOSPC4380-100G APOSPC4380-25G
5-Fluorouracil 99%
5-Fluorouracil

Antineoplastic agent. Not USP. Not for drug use.

Formula: C₄H₃FN₂O₂
MW: 130,08 g/mol
Boiling Pt: 190…200 °C/0,1mmHg
Melting Pt: 282…286 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006018
CAS Number: 51-21-8
EINECS: 200-085-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR