3-Fluoro-4-nitrobenzylalcohol 98%

Supplier: Apollo Scientific
Warning

Synonyms: (3-Fluoro-4-nitrophenyl)methanol

PC4403-5G PC4403-1G PC4403-10G
APOSPC4403-5GEA 143 EUR
APOSPC4403-5G APOSPC4403-1G APOSPC4403-10G
3-Fluoro-4-nitrobenzylalcohol 98%
3-Fluoro-4-nitrobenzylalkohol
Formula: C₇H₆FNO₃
MW: 171,1300 g/mol
Melting Pt: 93…97 °C
Storage Temperature: Ambient
MDL Number: MFCD03094236
CAS Number: 503315-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR