4-Chloro-5-fluoropyrimidine 98%

Supplier: Apollo Scientific
Warning

PC4448-1G PC4448-25G PC4448-5G
APOSPC4448-1GEA 102 EUR
APOSPC4448-1G APOSPC4448-25G APOSPC4448-5G
4-Chloro-5-fluoropyrimidine 98%
4-Chloro-5-fluoropyrimidine
Formula: C₄H₂ClFN₂
Storage Temperature: Refrigerator
MDL Number: MFCD09750157
CAS Number: 347418-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR