4-(Trifluoromethyl)benzamidine hydrochloride

Supplier: Apollo Scientific
Warning

PC446016-5G PC446016-1G
APOSPC446016-5GEA 30.1 EUR
APOSPC446016-5G APOSPC446016-1G
4-(Trifluoromethyl)benzamidine hydrochloride
4-(Trifluoromethyl)benzamidine hydrochloride
Formula: C₈H₇F₃N₂·HCl
Melting Pt: 167 °C
Storage Temperature: Ambient
MDL Number: MFCD04114431
CAS Number: 38980-96-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR