Trifluoropyruvyl amide
Supplier: Apollo Scientific
|
Warning
|
PC4476-1G
PC4476-250MG
PC4476-100MG
APOSPC4476-1GEA
1020
EUR
APOSPC4476-1G
APOSPC4476-250MG
APOSPC4476-100MG
Trifluoropyruvyl amide
Trifluoropyruvyl amide
Formula:
C₃H₂F₃NO₂ MW: 141,05 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04038369 CAS Number: 883500-27-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...