2-(5-Bromo-2-methylbenzyl)-5-(4-fluorophenyl)thiophene 98%

Supplier: Apollo Scientific

PC448035-5G
APOSPC448035-5GEA 81.2 EUR
APOSPC448035-5G
2-(5-Bromo-2-methylbenzyl)-5-(4-fluorophenyl)thiophene 98%
2-(5-Bromo-2-methylbenzyl)-5-(4-fluorophenyl)thiophene
Formula: C₁₈H₁₄BrFS
MW: 361,27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21496340
CAS Number: 1030825-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR