Heptafluorobutyryl chloride 97%

Supplier: Apollo Scientific
Danger

Synonyms: Perfluorobutyryl chloride, 2,2,3,3,4,4,4-Heptafluorobutyryl chloride

PC4490-25G PC4490-100G
APOSPC4490-25GEA 238 EUR
APOSPC4490-25G APOSPC4490-100G
Heptafluorobutyryl chloride 97%
Heptafluorobutyryl chloride
Formula: C₄ClF₇O
MW: 232,48 g/mol
Boiling Pt: 38…39 °C
Density: 1,556 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000707
CAS Number: 375-16-6
EINECS: 206-785-8
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR