1-[4-(Trifluoromethyl)phenyl]cyclobutanecarboxylic acid
Supplier: Apollo Scientific
PC449017-1G
PC449017-100MG
PC449017-250MG
PC449017-500MG
PC449017-5G
APOSPC449017-1GEA
401
EUR
APOSPC449017-1G
APOSPC449017-100MG
APOSPC449017-250MG
APOSPC449017-500MG
APOSPC449017-5G
1-[4-(Trifluoromethyl)phenyl]cyclobutanecarboxylic acid
1-[4-(Trifluoromethyl)phenyl]cyclobutanecarboxylic acid
Formula:
C₁₂H₁₁F₃O₂ MW: 244,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10703525 CAS Number: 1086379-78-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...