1-(2-Fluorophenyl)propanol

Supplier: Apollo Scientific

PC450219-10G PC450219-50G
APOSPC450219-10GEA 132 EUR
APOSPC450219-10G APOSPC450219-50G
1-(2-Fluorophenyl)propanol
1-(2-Fluorophenyl)propanol
Formula: C₉H₁₁FO
MW: 154.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08062368
CAS Number: 156022-15-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR