1H,1H-Perfluoroheptylamine 97%

Supplier: Apollo Scientific
Danger

Synonyms: 1H,1H-Tridecafluoroheptylamine

PC4503-1G PC4503-5G PC4503-25G
APOSPC4503-1GEA 89.9 EUR
APOSPC4503-1G APOSPC4503-5G APOSPC4503-25G
1H,1H-Perfluoroheptylamine 97%
1H,1H-Perfluoroheptylamine
Formula: C₇H₄F₁₃N
MW: 349,1 g/mol
Boiling Pt: 129 °C/740mmHg
Density: 1,624 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD01319145
CAS Number: 423-49-4
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR