Heptafluoropropyl-1,2,2,2-tetrafluoroethyl ether 97%
Supplier: Apollo Scientific
|
Warning
|
PC4513-100G
PC4513-25G
PC4513-5G
APOSPC4513-100GEA
662
EUR
APOSPC4513-100G
APOSPC4513-25G
APOSPC4513-5G
Heptafluoropropyl-1,2,2,2-tetrafluoroethyl ether 97%
Heptafluoropropyl-1,2,2,2-tetrafluoroethyl ether
Formula:
C₅HF₁₁O MW: 286,05 g/mol Boiling Pt: 40…42 °C Melting Pt: –175 °C Density: 1,538 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00042308 CAS Number: 3330-15-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...