2-Chloro-6-(trifluoromethyl)pyridine 98%
Supplier: Apollo Scientific
|
Danger
|
PC4544-25G
PC4544-100G
APOSPC4544-25GEA
21.9
EUR
APOSPC4544-25G
APOSPC4544-100G
2-Chloro-6-(trifluoromethyl)pyridine 98%
2-Chloro-6-(trifluoromethyl)pyridine
Formula:
C₆H₃ClF₃N MW: 181,54 g/mol Boiling Pt: 60…62 °C/8mmHg Melting Pt: 32…36 °C Flash Pt: 79,4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042226 CAS Number: 39890-95-4 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...