2-Amino-6-fluoropyridine 98%

Supplier: Apollo Scientific
Warning

PC4549-100G PC4549-25G PC4549-5G
APOSPC4549-100GEA 170 EUR
APOSPC4549-100G APOSPC4549-25G APOSPC4549-5G
2-Amino-6-fluoropyridine 98%
2-Amino-6-fluoropyridine
Formula: C₅H₅FN₂
MW: 112,11 g/mol
Melting Pt: 57…62 °C
Storage Temperature: Ambient
MDL Number: MFCD04114200
CAS Number: 1597-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR