2-(Trifluoromethyl)thiophene 97%
Supplier: Apollo Scientific
|
Warning
|
PC4594-1G
PC4594-250MG
APOSPC4594-1GEA
361
EUR
APOSPC4594-1G
APOSPC4594-250MG
2-(Trifluoromethyl)thiophene 97%
2-(Trifluoromethyl)thiophene
Formula:
C₅H₃F₃S Boiling Pt: 95 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD04973785 CAS Number: 86093-76-7 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...