(4-Fluorophenyl)methanesulphonyl chloride 95%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
4-Fluoro-α-toluenesulphonyl chloride
PC4605-1G
PC4605-5G
APOSPC4605-1GEA
96
EUR
APOSPC4605-1G
APOSPC4605-5G
(4-Fluorophenyl)methanesulphonyl chloride 95%
(4-Fluorophenyl)methanesulphonyl chloride
Formula:
C₇H₆ClFO₂S MW: 208,64 g/mol Melting Pt: 60…63 °C Flash Pt: 107 °C Storage Temperature: Ambient |
MDL Number:
MFCD01631930 CAS Number: 103360-04-9 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...