4-Ethoxy-2-fluoro-1-nitrobenzene 98%
Supplier: Apollo Scientific
|
Warning
|
PC4607-1G
PC4607-5G
PC4607-25G
APOSPC4607-1GEA
132
EUR
APOSPC4607-1G
APOSPC4607-5G
APOSPC4607-25G
4-Ethoxy-2-fluoro-1-nitrobenzene 98%
4-Ethoxy-2-fluoro-1-nitrobenzene
Formula:
C₈H₈FNO₃ Melting Pt: 47…49 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD04115633 CAS Number: 28987-48-6 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...