1,1,1,2,3,4,4,4-Octafluorobutane 95%

Supplier: Apollo Scientific
Warning

PC4617-25G PC4617-5G
APOSPC4617-25GEA 870 EUR
APOSPC4617-25G APOSPC4617-5G
1,1,1,2,3,4,4,4-Octafluorobutane 95%
1,1,1,2,3,4,4,4-Octafluorobutane
Formula: C₄H₂F₈
MW: 202.05 g/mol
Boiling Pt: 33…34 °C
Storage Temperature: Ambient
MDL Number: MFCD03092909
CAS Number: 75995-72-1
UN: 3163
ADR: 2.2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR