5-Fluoro-2-methylphenylmagnesium bromide
Supplier: Apollo Scientific
PC4644-100ML
PC4644-50ML
APOSPC4644-100MLEA
531
EUR
APOSPC4644-100ML
APOSPC4644-50ML
5-Fluoro-2-methylphenylmagnesium bromide
5-Fluoro-2-methylphenylmagnesium bromide
Formula:
C₇H₆BrFMg Boiling Pt: 65 °C Density: 0,96 g/cm³ (25 °C) Flash Pt: –17 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01311468 CAS Number: 186496-59-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...