2,2,3,4,4,4-Hexafluoro-1-butanol 95%

Supplier: Apollo Scientific
Warning

PC4653-100G PC4653-25G PC4653-500G
APOSPC4653-100GEA 41.6 EUR
APOSPC4653-100G APOSPC4653-25G APOSPC4653-500G
2,2,3,4,4,4-Hexafluoro-1-butanol 95%
2,2,3,4,4,4-Hexafluoro-1-butanol

Contains up to 5% CF3CF(CH2OH)CHF2

Formula: C₄H₄F₆O
Boiling Pt: 114…115 °C
Density: 1,564 g/cm³ (25 °C)
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00042309
CAS Number: 382-31-0
EINECS: 206-842-7
UN: 1987
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR