Hexafluoroglutaramide 97%

Supplier: Apollo Scientific
Warning

PC4690-5G PC4690-1G
APOSPC4690-5GEA 212 EUR
APOSPC4690-5G APOSPC4690-1G
Hexafluoroglutaramide 97%
Hexafluoroglutaramide
Formula: C₅H₄F₆N₂O₂
MW: 238.10 g/mol
Melting Pt: 209…211 °C
Storage Temperature: Ambient
MDL Number: MFCD00014804
CAS Number: 507-68-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR