N,N-Diethyl(2-chloro-1,1,2-trifluoroethyl)amine 90%
Supplier: Apollo Scientific
|
Danger
|
PC4728-100G
PC4728-25G
PC4728-5G
APOSPC4728-100GEA
815
EUR
APOSPC4728-100G
APOSPC4728-25G
APOSPC4728-5G
N,N-Diethyl(2-chloro-1,1,2-trifluoroethyl)amine 90%
N,N-Diethyl(2-chloro-1,1,2-trifluoroethyl)amine
Formula:
C₆H₁₁ClF₃N Boiling Pt: 38 °C/10mmHg Density: 1,19 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00054672 CAS Number: 357-83-5 UN: 3267 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...