Bis(1H,1H-perfluorobutyl)fumarate 97%

Supplier: Apollo Scientific

PC4782-25G PC4782-5G PC4782-1G
APOSPC4782-25GEA 728 EUR
APOSPC4782-25G APOSPC4782-5G APOSPC4782-1G
Bis(1H,1H-perfluorobutyl)fumarate 97%
Bis(1H,1H-perfluorobutyl)fumarate
Formula: C₁₂H₆F₁₄O₄
MW: 480,16 g/mol
Melting Pt: 40…42 °C
Storage Temperature: Ambient
MDL Number: MFCD00337741
CAS Number: 24120-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR