Perfluoropropene 99%

Supplier: Apollo Scientific
Warning

Synonyms: Hexafluoropropene

PC4790-100G PC4790-500G
APOSPC4790-100GEA 219 EUR
APOSPC4790-100G APOSPC4790-500G
Perfluoropropene 99%
Perfluoropropene
Formula: C₃F₆
MW: 150,02 g/mol
Boiling Pt: –29 °C
Melting Pt: –153 °C
Density: 1,583 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000447
CAS Number: 116-15-4
EINECS: 204-127-4
UN: 1858
ADR: 2.2,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR