3-Amino-2-bromo-5-fluoropyridine 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Bromo-5-fluoropyridin-3-amine, 2-Bromo-5-fluoro-3-pyridylamine
PC48312-10G
PC48312-25G
PC48312-5G
APOSPC48312-10GEA
129
EUR
APOSPC48312-10G
APOSPC48312-25G
APOSPC48312-5G
3-Amino-2-bromo-5-fluoropyridine 98%
3-Amino-2-bromo-5-fluoropyridine
Formula:
C₅H₄BrFN₂ Melting Pt: 97…101 °C Storage Temperature: Ambient |
MDL Number:
MFCD05662413 CAS Number: 884495-03-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...