5-Bromo-2-hydroxy-3-(trifluoromethyl)pyridine 98%

Supplier: Apollo Scientific
Warning

Synonyms: 5-Bromo-3-(trifluoromethyl)pyridin-2-ol

PC48328-25G
APOSPC48328-25GEA 244 EUR
APOSPC48328-25G
5-Bromo-2-hydroxy-3-(trifluoromethyl)pyridine 98%
5-Bromo-2-hydroxy-3-(trifluoromethyl)pyridine
Formula: C₆H₃BrF₃NO
MW: 242,00 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12546486
CAS Number: 76041-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR