2-Bromo-5-(trifluoromethyl)benzoic acid 97%

Supplier: Apollo Scientific
Warning

PC48364-100G PC48364-25G PC48364-5G
APOSPC48364-100GEA 495 EUR
APOSPC48364-100G APOSPC48364-25G APOSPC48364-5G
2-Bromo-5-(trifluoromethyl)benzoic acid 97%
2-Bromo-5-(trifluoromethyl)benzoic acid
Formula: C₈H₄BrF₃O₂
MW: 269,0154 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06208253
CAS Number: 1483-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR