5'-Fluoro-2'-iodoacetophenone 98%

Supplier: Apollo Scientific

PC48743-10G PC48743-5G
APOSPC48743-10GEA 80 EUR
APOSPC48743-10G APOSPC48743-5G
5'-Fluoro-2'-iodoacetophenone 98%
5'-Fluoro-2'-iodoacetophenone
Formula: C₈H₆FIO
Storage Temperature: Ambient
MDL Number: MFCD09033159
CAS Number: 914225-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR