3-Amino-6-(trifluoromethyl)-1H-indazole

Supplier: Apollo Scientific
Warning

Synonyms: 6-(Trifluoromethyl)-1H-indazol-3-amine

PC49001-1G PC49001-250MG
APOSPC49001-1GEA 58 EUR
APOSPC49001-1G APOSPC49001-250MG
3-Amino-6-(trifluoromethyl)-1H-indazole
3-Amino-6-(trifluoromethyl)-1H-indazole
Formula: C₈H₆F₃N₃
MW: 201,148 g/mol
Melting Pt: 163…165 °C
Storage Temperature: Ambient
MDL Number: MFCD03426695
CAS Number: 2250-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR