Tri-tert-butylphosphonium tetrafluoroborate 96%

Supplier: Apollo Scientific
Warning

Synonyms: Tri-t-butylphosphonium tetrafluoroborate

PC49062-100G PC49062-25G PC49062-5G
APOSPC49062-100GEA 263 EUR
APOSPC49062-100G APOSPC49062-25G APOSPC49062-5G
Tri-tert-butylphosphonium tetrafluoroborate 96%
Tri-tert-butylphosphonium tetrafluoroborate
Formula: C₁₂H₂₈BF₄P
MW: 290,13 g/mol
Melting Pt: 260…261 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD04039975
CAS Number: 131274-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR