2-Bromo-6-chlorobenzotrifluoride 97%

Supplier: Apollo Scientific
Warning

Synonyms: 1-Bromo-3-chloro-2-(trifluoromethyl)benzene

PC49063-10G PC49063-25G PC49063-5G
APOSPC49063-10GEA 241 EUR
APOSPC49063-10G APOSPC49063-25G APOSPC49063-5G
2-Bromo-6-chlorobenzotrifluoride 97%
2-Bromo-6-chlorobenzotrifluoride
Formula: C₇H₃BrClF₃
MW: 259,4500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08459291
CAS Number: 857061-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR