2-(6-Chloro-3-pyridazinyl)-2-(2-fluorophenyl)acetonitrile 97%

Supplier: Apollo Scientific
Warning

PC4910-1G PC4910-5G PC4910-10G
APOSPC4910-1GEA 132 EUR
APOSPC4910-1G APOSPC4910-5G APOSPC4910-10G
2-(6-Chloro-3-pyridazinyl)-2-(2-fluorophenyl)acetonitrile 97%
2-(6-Chloro-3-pyridazinyl)-2-(2-fluorophenyl)acetonitrile
Formula: C₁₂H₇ClFN₃
Melting Pt: 69…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00202113
CAS Number: 154419-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR