3-(Difluoromethyl)benzoic acid

Supplier: Apollo Scientific
Warning

PC49197-1G PC49197-250MG PC49197-5G
APOSPC49197-1GEA 111 EUR
APOSPC49197-1G APOSPC49197-250MG APOSPC49197-5G
3-(Difluoromethyl)benzoic acid
3-(Difluoromethyl)benzoic acid
Formula: C₈H₆F₂O₂
Melting Pt: 127…129 °C
Storage Temperature: Ambient
MDL Number: MFCD17480442
CAS Number: 55805-20-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR